N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H66982.06
also known as H66982-06
5 g
Catalog number H66982.06
also known as H66982-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS86061-01-0
IUPAC Name1-tert-butyl 2,3,4,5,6-pentafluorophenyl 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanedioate
Molecular FormulaC29H24F5NO6
InChI KeyDWYWJUBBXKYAMY-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
FormPowder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only