Ethyl 5-nitroindole-2-carboxylate, 97%
Ethyl 5-nitroindole-2-carboxylate, 97%
Ethyl 5-nitroindole-2-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 5-nitroindole-2-carboxylate, 97%

CAS: 16732-57-3 | C11H10N2O4 | 234.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66997.03
also known as H66997-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS16732-57-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormPowder
Assay from Supplier's CofA≥96.0%
Assay (unspecified)Water: ≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only