Maybridge HitCreator Screening Library
Maybridge™
Maybridge HitCreator Screening Library
13,800 compounds conveniently supplied as dry films in Thermo Scientific Matrix 96 shallow-well plates
Have Questions?
Change viewbuttonViewtableView
Catalog NumberKit Contents
HC02038445 Plates, 0.25 μmol
HC020000180 Plates, 1 μmol
Catalog number HC020384
Price (USD)
-
Kit Contents:
45 Plates, 0.25 μmol
13,800 compounds conveniently supplied as dry films in Thermo Scientific Matrix 96 shallow-well plates
HitCreator is derived from a rigorous analysis of more than 550,000 commercially available screening compounds to offer exceptional diversity for such a small library
RUO – Research Use Only
Specifications
For Use With (Application)High Throughput Screening
Kit Contents45 Plates, 0.25 μmol
Plate MaterialMatrix plate
Product TypeScreening Compound Library
Unit Size-