Please note: The color of SureCast Sealing Pads has changed from black to grey.
SureCast™ Gel Handcast Bundle B - Hardware Only
Invitrogen™
SureCast™ Gel Handcast Bundle B - Hardware Only
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle B. The complete product includes theRead more
Have Questions?
Catalog NumberQuantity
HC1000S1 kit
Catalog number HC1000S
Price (USD)
536.00
Each
Quantity:
1 kit
Price (USD)
536.00
Each
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle B. The complete product includes the SureCast™ Handcast System. For a bundle that also includes the SureCast™ Handcast Reagents, please see SureCast™ Gel Handcast Bundle A. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.
Features of the SureCast Handcast System include:
  • Innovative, leak-free design—minimizes the number of failed gels, resulting in less wasted time
  • Superior glass plate durability
  • Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
  • Unique tilt feature for easy loading of acrylamide solutions
  • The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
The SureCast Handcast Bundle B includes all the hardware you need to get started:
  • 2 SureCast Handcast Stations
  • 2 SureCast™ Glass Plate Sets (2 front and 2 back)
  • 6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
  • 10 SureCast™ Gel Spacers
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Unit SizeEach
Frequently asked questions (FAQs)
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.