Product Image
Invitrogen™
SureCast™ Multi-Use Tool, 10-well
The SureCast™ Multi-Use Tool is a gel knife, gel releaser, and well template guide all in one convenient tool. DesignedRead more
Have Questions?
Catalog NumberQuantity
HC10101 unit
Catalog number HC1010
Price (USD)
31.42
Each
Quantity:
1 unit
Price (USD)
31.42
Each
The SureCast™ Multi-Use Tool is a gel knife, gel releaser, and well template guide all in one convenient tool. Designed for use with the SureCast™ Handcast System, this innovative guide outlines the wells for easy visualization and loading of samples into the gel. After electrophoresis, the tool also allows you to easily separate the two glass plates, cut the gel foot, and release the gel.
Includes:
1 SureCast 10-well Multi-Use Tool
Related products:
SureCast™ Handcast Station
SureCast™ Glass Plate Set
SureCast™ Gel Spacers
SureCast™ Sealing Pads
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 unit
Shipping ConditionRoom Temperature
For Use With (Equipment)SureCast Vertical Gel System
Gel SizeMini
Product LineSureCast
TypeSureCast™ Multi-use Tool, 10-well
Unit SizeEach
Frequently asked questions (FAQs)
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.