SureCast™ Multi-Use Tool, 15-well - FAQs
View additional product information for SureCast™ Multi-Use Tool, 15-well - FAQs (HC1015)
3 product FAQs found
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.