HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 208-230V/60Hz, 6.5A - FAQs
HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 12.7 cu. ft. (359.6L) capacity, 208-230V/60Hz, 6.5A - FAQs
View additional product information for HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (HFC1350TD)