HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 20 cu. ft. (566.3L) capacity, 115V/60Hz, 6.5A - FAQs
HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 20 cu. ft. (566.3L) capacity, 115V/60Hz, 6.5A - FAQs
View additional product information for HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (HFC2050TA)