HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 115V/60Hz, 5A - FAQs
HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 115V/60Hz, 5A - FAQs
View additional product information for HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezers - FAQs (HFC350TA)