HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 14.9 cu. ft. (420.9L) capacity, holds 300 boxes, 115V/60Hz - FAQs
HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 14.9 cu. ft. (420.9L) capacity, holds 300 boxes, 115V/60Hz - FAQs
View additional product information for HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (HFU300TA)