HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes, 115V/60Hz, 16A, painted galvaneel interior - FAQs
HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezer, 23 cu. ft. (651.3L) capacity, holds 400 boxes, 115V/60Hz, 16A, painted galvaneel interior - FAQs
View additional product information for HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (HFU400BA)