HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes, 208-230V/60Hz, 12A - FAQs
HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes, 208-230V/60Hz, 12A - FAQs
View additional product information for HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (HFU40240BD)