HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capacity, holds 320 boxes, 115V/60Hz, 16A - FAQs
HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezer, 17.3 cu. ft. (489.9L) capacity, holds 320 boxes, 115V/60Hz, 16A - FAQs
View additional product information for HERAfreeze™ HFU B Series -40°C Upright Ultra-Low Temperature Freezers - FAQs (HFU40320BA)