HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 24.1 cu. ft. (681.5L) capacity, holds 500 boxes, 115V/60Hz - FAQs
HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 24.1 cu. ft. (681.5L) capacity, holds 500 boxes, 115V/60Hz - FAQs
View additional product information for HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (HFU500TA)