HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezer, 28 cu. ft. (792.8L) capacity, holds 600 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezer, 28 cu. ft. (792.8L) capacity, holds 600 boxes, 208-230V/60Hz, 12A, painted galvaneel interior - FAQs
View additional product information for HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (HFU600BD)