HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 115V/60Hz - FAQs
HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezer, 28.8 cu. ft. (815.1L) capacity, holds 600 boxes, 115V/60Hz - FAQs
View additional product information for HERAfreeze™ HFU T Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (HFU600TA)