Sample Storage Starter Packs
Thermo Scientific™
Sample Storage Starter Packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
HHD-WP1D-2Tubes and Decapper
HHD-ST-WP1D-2Tubes, Decapper, ST Reader
HHD-HSX-WP1D-2Tubes, Decapper, HSX Reader
HHD-HSX-WP1D-5050.5mL Tubes, Decapper, Reader
HHD-HSX-WP1D-5101.0mL Tubes, Decapper, Reader
Catalog number HHD-WP1D-2
Price (USD)
2,666.00
Case of 1
Includes:
Tubes and Decapper
Price (USD)
2,666.00
Case of 1
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
  • Save 40% compared to buying the products individually.
  • Choose from five different starter pack options.
  • Eliminate the need to reformat non-2D cryotube samples.
  • Secure sample tracking with 2D, 1D, and human readable codes.
  • Cap/decap a full rack of tubes in less than 1 minute.
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Basic Starter Pack Includes:
  • 8-Channel Screw Cap Decapper (1 each)
  • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
  • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Advanced Starter Pack Includes:
  • 8-Channel Screw Cap Decapper (1 each)
  • VisionMate ST Single-Tube 2D Barcode Reader (1 each)
  • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
  • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium Starter Pack Includes:
  • 8-Channel Screw Cap Decapper (1 each)
  • VisionMate HSX High Speed Barcode Reader (1 each)
  • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
  • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium 0.5 mL Starter Pack Includes:
  • 8-Channel Screw Cap Decapper (1 each)
  • VisionMate HSX High Speed Barcode Reader (1 each)
  • 0.5 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Premium 1.0 mL Starter Pack Includes:
  • 8-Channel Screw Cap Decapper (1 each)
  • VisionMate HSX High Speed Barcode Reader (1 each)
  • 1.0 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Specifications
IncludesTubes and Decapper
TypeBasic Starter Pack
Unit SizeCase of 1