Human Suspension Hepatocytes, Metabolism Qualified, Female, (4-8 x 10<sup>6</sup> cells)
Gibco™

Human Suspension Hepatocytes, Metabolism Qualified, Female, (4-8 x 106 cells)

These cryopreserved human female hepatocytes are suitable for use in suspension metabolism applications.Cells per vial: 4–8 millionThe commonly tested PhaseRead more
Have Questions?
Catalog NumberQuantity
HMCS2S1 vial
Catalog number HMCS2S
Price (USD)
739.65
Online exclusive
1,000.00
Save 260.35
Each
Quantity:
1 vial
Price (USD)
739.65
Online exclusive
1,000.00
Save 260.35
Each
These cryopreserved human female hepatocytes are suitable for use in suspension metabolism applications.

Cells per vial: 4–8 million

The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes were analyzed for metabolic activity in each lot. Metabolite formation was measured by standard biochemical assays using LC/MS/MS or HPLC.

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
• Phase I and II enzyme activities
• Proper morphology
• Viabilities generally >75%

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links:

Learn more about our cryopreserved hepatocytes

Learn about our full line of hepatocytes ,

Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor SourceSingle Donor
No. of Cells4-8 x 106
Product LineGibco
Product TypeHepatocytes
Quantity1 vial
Tested ForPhase I and II Enzyme Activity, Proper Morphology, Viability Generally >75%
Cell TypeHepatocytes
Culture TypeSuspension Cell Culture
FormCryopreserved
GenderFemale
SpeciesHuman
Unit SizeEach
Contents & Storage
Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.

Frequently asked questions (FAQs)

Who should I contact if I am looking for hepatocytes with particular characteristics?

Our hepatocytes are prequalified and sold according to application, such as induction, short-term metabolism, and transporter uptake. If you require hepatocytes with particular P450 values or donor specifications, contact our Hepatic Biology Tech Support Team (hepaticproducts@thermofisher.com.com) or (866) 952-3559 to help with specific lot recommendations.

Who can I call if I have technical questions about hepatocyte cell culture?

Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocyte cell culture. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952-3559

Are there guidelines for the procurement of human liver tissue?

Thermofisher Scientific works with a variety of human tissue sources, including Tissue and Organ Procurement Organizations, qualified Research Tissue Organizations and prominent academic and medical centers through collaborations that follow rigorous regulations, certifications and/or accreditations (“Source Facilities”). Tissues obtained through source facilities are consistent with the applicable legal and ethical practices of the United States.

Source Facilities are required by federal law to maintain accurate records and donor confidentiality as well as adhere to certain applicable laws governing tissue used for research globally. These include, but are not limited to:

- 45 CFR Part 46, subparts A, B, C, D where applicable (US federal guidance on the use of human subjects in research)
- Health Insurance Portability and Accountability Act (HIPAA) (addressing issues of confidentiality of personal information)
- National Organ Transplant Act (42 CFR Part 482)
- The Uniform Anatomical Gift Act (UAGA) of 1968, revised 1984

Where do you get your human liver tissue?

The human liver cells are derived from tissue obtained from accredited institutions. Consent is obtained by these institutions from the donor or the donor’s legal next of kin, for the use of the tissue and its derivatives for research purpose.

What should I plate hepatocyte cells on?

Hepatocytes need to be plated on collagen I-coated culture vessels.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.