HEP50 Pooled Human Cryopreserved Hepatocytes
Gibco™

HEP50 Pooled Human Cryopreserved Hepatocytes

Get the convenience, affordability, and power of a pooled population of hepatocytes in a single vial. HEP50 Pooled Human CryopreservedRead more
Have Questions?
Catalog NumberDonor Source
HMCS50Pooled
Catalog number HMCS50
Price (ZAR)
-
Donor Source:
Pooled
Get the convenience, affordability, and power of a pooled population of hepatocytes in a single vial. HEP50 Pooled Human Cryopreserved Hepatocytes are a representative population of 50 different donor hepatocytes that have been isolated using our novel proprietary CellStream Isolation Technology, helping to deliver consistent, predictable results in your drug metabolism studies and other biological assays.

• Pure, highly viable hepatocytes isolated using our CellStream Isolation Technology
• 50 donor hepatocytes, 25 male and 25 female, in each vial
• Verified enzymatic and pathway activity

Each lot is tested for:
• Viability ≥ 75%
• Phase I and II enzyme activities
• Transporter suspension uptake

CellStream Isolation Technology
HEP50 Pooled Human Hepatocytes are produced using our novel proprietary CellStream Isolation Technology. This process eliminates the disadvantages of density gradient fractionation prior to cryopreservation of the pool. Instead, we gently isolate and purify the pre-screened, high-viability, and metabolically-active hepatocytes from other cells and debris. The result is a remarkably pure pooled hepatocyte preparation with high viability for use in drug metabolism studies and other biological assays.

50 donors in one vial
Each vial of HEP50 Pooled Human Hepatocytes contains hepatocytes from 50 individual donors: 25 female and 25 male.

Verified activity
Each pool is characterized for CYP1A2, CYP2B6, CYP2C8, CYP2C9, CYP2C19, CYP2D6, CYP2E1, CYP3A4, CYP3A4/5, and FMO activity, as well as NTCP, OATP1B3, and OATP transporter pathways.

Ready for your drug metabolism experiments
HEP50 Pooled Human Hepatocytes are useful in a wide variety of applications, including intrinsic clearance, transport, toxicology, biochemistry, and cell biology studies.

Note: These products require shipment in liquid nitrogen vapor dewars, so additional fees may apply.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor SourcePooled
Product TypeHepatocytes
AgeAdult
Cell TypeHepatocytes
FormFrozen
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in a cryostorage dewar or -135°C freezer.

Frequently asked questions (FAQs)

Who can I call if I have technical questions regarding cryopreserved hepatocytes?

Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocytes. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952 3559

Who should I contact if I am looking for hepatocytes with particular characteristics?

Our hepatocytes are prequalified and sold according to application, such as induction, short-term metabolism, and transporter uptake. If you require hepatocytes with particular P450 values or donor specifications, contact our Hepatic Biology Tech Support Team (hepaticproducts@thermofisher.com.com) or (866) 952-3559 to help with specific lot recommendations.

Who can I call if I have technical questions about hepatocyte cell culture?

Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocyte cell culture. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952-3559

Are there guidelines for the procurement of human liver tissue?

Thermofisher Scientific works with a variety of human tissue sources, including Tissue and Organ Procurement Organizations, qualified Research Tissue Organizations and prominent academic and medical centers through collaborations that follow rigorous regulations, certifications and/or accreditations (“Source Facilities”). Tissues obtained through source facilities are consistent with the applicable legal and ethical practices of the United States.

Source Facilities are required by federal law to maintain accurate records and donor confidentiality as well as adhere to certain applicable laws governing tissue used for research globally. These include, but are not limited to:

- 45 CFR Part 46, subparts A, B, C, D where applicable (US federal guidance on the use of human subjects in research)
- Health Insurance Portability and Accountability Act (HIPAA) (addressing issues of confidentiality of personal information)
- National Organ Transplant Act (42 CFR Part 482)
- The Uniform Anatomical Gift Act (UAGA) of 1968, revised 1984

Where do you get your human liver tissue?

The human liver cells are derived from tissue obtained from accredited institutions. Consent is obtained by these institutions from the donor or the donor’s legal next of kin, for the use of the tissue and its derivatives for research purpose.