Human Suspension Hepatocytes, Polymorphic Donors, (4-8 x 106 cells)

Catalog number: HMCSPS

Gibco™  Related applications: Hepatocytes & Hepatic Models | Pharma & Biopharma | Primary Cell Culture | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Each lot of cryopreserved human hepatocytes is suitable for use in suspension metabolism applications and pharmacogenetically has been characterized as an homozygous null allele (e.g. CYP2D6*4⁄*4) via genotyping analysis (TaqMan®). The commonly tested Phase I (P450) and Phase II (UGT, SULT) drug metabolizing enzymes are analyzed for metabolic activity in each lot. Metabolite formation is measured by standard biochemical assays using LC-MS/MS or HPLC.

Cells per vial: 4-8 million

Get the Results You Need with GIBCO® Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro/in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:

•Phase I and II enzyme activities
•Proper morphology
•Viabilities generally >75%

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links

Learn more about our cryopreserved hepatocytes

Learn about our full line of hepatocytes,

Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Passage Number: Passage 0
Cell Type: Hepatocytes
Culture Type: Suspension Cell Culture
Donor Source: Single Donor
Form: Cryopreserved
Format: Tube(s)
Number of Cells: 4-8 x 106
Product Line: Gibco®
Product Size: 1 vial
Species: Human

Contents & storage

Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.

Documents

Manuals & protocols