Human Suspension Hepatocytes, Transporter Qualified, (9-12 x 106 cells) - FAQs
Human Suspension Hepatocytes, Transporter Qualified, (9-12 x 106 cells) - FAQs
View additional product information for Human Suspension Hepatocytes, Transporter Qualified, (9-12 x 106 cells) - FAQs (HMCSTL)
1 product FAQs found
Will I be able to transfect my primary hepatocyte cells if I use an overlay?
No. The overlay will interfere with transfection. For transfection assays, we recommend that you perform the transfection first, and then proceed to overlay the cells.