Product Image
Gibco™

Human S9 Fractions

Human S9 Fractions
Have Questions?
Catalog NumberQuantity
HMS9PL1 mL
Catalog number HMS9PL
Price (USD)
127.65
Online exclusive
173.00
Save 45.35
Each
Quantity:
1 mL
Price (USD)
127.65
Online exclusive
173.00
Save 45.35
Each

Pooled human liver S9 fractions.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
AgeAdult
Concentration20 mg/mL
Donor SourcePooled
GenderMixed Gender
Quantity1 mL
Shipping ConditionDry Ice
Sub Cellular LocalizationS9 Fraction
Target SpeciesHuman
TissueLiver
FormFrozen
Product LineGibco
Product TypeSubcellular Fractions
Unit SizeEach
Contents & Storage
Contains a single 1 mL tube at a concentration of 20 mg/mL. Store between -60°C and -85°C.

Frequently asked questions (FAQs)

Who should I contact if I am looking for hepatocytes with particular characteristics?

Our hepatocytes are prequalified and sold according to application, such as induction, short-term metabolism, and transporter uptake. If you require hepatocytes with particular P450 values or donor specifications, contact our Hepatic Biology Tech Support Team (hepaticproducts@thermofisher.com.com) or (866) 952-3559 to help with specific lot recommendations.

Who can I call if I have technical questions about hepatocyte cell culture?

Thermo Fisher Scientific has a dedicated team to answer questions regarding hepatocyte cell culture. You can send a question by email (hepaticproducts@thermofisher.com) or contact us by phone: US toll-free: (866) 952-3559

Are there guidelines for the procurement of human liver tissue?

Thermofisher Scientific works with a variety of human tissue sources, including Tissue and Organ Procurement Organizations, qualified Research Tissue Organizations and prominent academic and medical centers through collaborations that follow rigorous regulations, certifications and/or accreditations (“Source Facilities”). Tissues obtained through source facilities are consistent with the applicable legal and ethical practices of the United States.

Source Facilities are required by federal law to maintain accurate records and donor confidentiality as well as adhere to certain applicable laws governing tissue used for research globally. These include, but are not limited to:

- 45 CFR Part 46, subparts A, B, C, D where applicable (US federal guidance on the use of human subjects in research)
- Health Insurance Portability and Accountability Act (HIPAA) (addressing issues of confidentiality of personal information)
- National Organ Transplant Act (42 CFR Part 482)
- The Uniform Anatomical Gift Act (UAGA) of 1968, revised 1984

Where do you get your human liver tissue?

The human liver cells are derived from tissue obtained from accredited institutions. Consent is obtained by these institutions from the donor or the donor’s legal next of kin, for the use of the tissue and its derivatives for research purpose.