PEPotec™ SRM Custom Peptide Libraries
Thermo Scientific™

PEPotec™ SRM Custom Peptide Libraries

Thermo Scientific PEPotec SRM Peptide Libraries are fully synthetic custom libraries for the development of mid- to high-throughput absolute quantitationRead more
Have Questions?
Catalog NumberQuantity
HPEPOMGR31 each
Catalog number HPEPOMGR3
Price (USD)
-
Quantity:
1 each
Thermo Scientific PEPotec SRM Peptide Libraries are fully synthetic custom libraries for the development of mid- to high-throughput absolute quantitation with selective-reaction monitoring (SRM) or multiple-reaction monitoring (MRM) assays. The PEPotec SRM Peptide Libraries offer great convenience and flexibility for the development of quantitative mass spectroscopy (MS) with many customizable options. The standard service supplies a suspension of at least 0.1 mg of each crude peptide housed in individual tubes in a 96-well plate format with either arginine (R) or lysine (K) as the C-terminal amino acid (other C-terminal amino acids are available). Three quality control grades are available, and optional services and peptide modifications are offered to give you the peptide libraries that fit your experimental needs.

Learn more about our PEPotec SRM Custom Peptide Libraries ›

Highlights:
• Traceable—peptides are provided in individual 2-D barcoded tubes in 96-tube plates
• Customized—libraries available in various grades with optional services available
• Convenient—standard libraries are delivered solubilized in 0.1% trifluoroacetic acid (TFA) in 50% (v/v) acetonitrile/water
• Flexible—extensive list of available modifications

Applications:
• Mid- to high-throughput development of SRM and MRM assays
• MS workflows with relative and absolute quantitation strategies

Includes:
• Fully synthetic, crude (as synthesized) peptides
• Multiple grades of QC analysis and optional services and modifications
• Provided in individual Thermo Scientific Matrix 96-tube plates

For a quote or more information, email us at: peps@thermofisher.com.
For Europe, Middle East, and Africa, please email us at: europeservicespeptides@thermofisher.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 each
Shipping ConditionWet Ice
Product LinePEPotec
Unit SizeEach
Contents & Storage
Store at -20°C upon receipt.