Screening Library Compounds, Hand-selected 96-well plate
Maybridge™

Screening Library Compounds, Hand-selected 96-well plate

Hand-selected screening compound library for your high throughput screening applications.
Have Questions?
Catalog NumberPlate Material
HTSPLATEMatrix plate
Catalog number HTSPLATE
Price (USD)
-
Plate Material:
Matrix plate
Customized selection of 50 compounds from within the Maybridge Screening Collection.
The Maybridge Screening collection consists of over 53,000 organic compounds, individually designed based on over 50 years of experience in heterocyclic chemistry.
RUO – Research Use Only
Specifications
For Use With (Application)High Throughput Screening
Plate MaterialMatrix plate
Product TypeScreening Compound Library
Unit Size-