Image-iT™ Low Density Lipoprotein Uptake Kit, pHrodo™ Red - FAQs
View additional product information for Image-iT™ Low Density Lipoprotein Uptake Kit, pHrodo™ Red - FAQs (I34360)
3 product FAQs found
Is the LDL in pHrodo Red-LDL acetylated or oxidized?
This product does not contain acetylated or oxidized LDL.
Why is fixation not recommended for cells stained with pHrodo Red-LDL?
pHrodo Red and Green dyes are pH-sensitive dyes. They should only be used on live cells to detect pH of cellular compartments. Upon fixation of the cells, the cell membrane is compromised and you are only measuring the pH of the buffer the samples are held in.
How is the pHrodo Red dye bound to the LDL in pHrodo Red-LDL?
pHrodo Red dye is covalently bound to the LDL.