Image-iT® FX Kit #1 (with Alexa Fluor® 350 Goat Anti-Mouse IgG, ProLong® Gold Antifade, and Image-iT® FX Signal Enhancer) - FAQs
Image-iT® FX Kit #1 (with Alexa Fluor® 350 Goat Anti-Mouse IgG, ProLong® Gold Antifade, and Image-iT® FX Signal Enhancer) - FAQs
View additional product information for Image-iT® FX Kit #1 (with Alexa Fluor® 350 Goat Anti-Mouse IgG, ProLong® Gold Antifade, and Image-iT® FX Signal Enhancer) - FAQs (I37150)
1 product FAQs found
I used Image-iT FX Signal Enhancer solution to get rid of nonspecific nuclear labeling with Alexa Fluor 568 secondary antibody, but I also saw a significant reduction in my specific mitochondrial antibody labeling. Why is this and what can I do?
The Image-iT FX Signal Enhancer reduces non-specific binding of dye conjugates by blocking positively-charged areas of cells or tissues that attract negatively-charged dyes. In cells after fixation, some positively-charged structures are the nuclei and mitochondria. Thus, you would expect to see a reduction in both mitochondrial and nuclear signal. The lower signal you see afterward is the specific mitochondrial signal; the fluorescence that was lost was the non-specific labeling.