Image-iT™ Cell Painting Kit - FAQs
Image-iT™ Cell Painting Kit - FAQs
View additional product information for Image-iT™ Cell Painting Kit - FAQs (I64000)
3 product FAQs found
Can I use the reagents in the Image-iT Cell Painting Kit with bacterial or fungal samples?
Yes, the reagents in the Image-iT Cell Painting Kit can be used with bacterial or fungal samples but the final working concentrations and incubation times would have to be optimized by the user.
Can I use the reagents in the Image-iT Cell Painting Kit with non-mammalian cell lines?
Yes, the reagents in the Image-iT Cell Painting Kit can be used with non-mammalian cell lines but the final working concentrations and incubation times would have to be optimized by the user.
When I order the Image-iT Cell Painting Kit, can I swap out some components of the kit to get them in a different fluorescent color?
Unfortunately, this is not possible. You may purchase alternative products with different fluorescent colors as standalone items.