Isotemp™ -86°C Ultra-Low Temperature Chest Freezers, 20 cu. ft., Capacity: 20 cu. ft. (566.3L); 208-230V/60Hz 12A - FAQs
Isotemp™ -86°C Ultra-Low Temperature Chest Freezers, 20 cu. ft., Capacity: 20 cu. ft. (566.3L); 208-230V/60Hz 12A - FAQs
View additional product information for Isotemp™ -86°C Ultra-Low Temperature Chest Freezers, 20 cu. ft. - FAQs (I920CD)