Pilas de transferencia iBlot™ 2, nitrocelulosa, tamaño normal
Invitrogen™

Pilas de transferencia iBlot™ 2, nitrocelulosa, tamaño normal

Se utiliza para la transferencia de proteínas con el dispositivo de transferencia de gel iBlot 2.
Have Questions?
Número de catálogoCantidad
IB2300110 pileup
Número de catálogo IB23001
Precio (MXN)
-
Cantidad:
10 pileup

iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This regular-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels or E-PAGE 48 or 96 gels.

Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Cantidad10 pileup
Dimensions (LxW)Normal, 13,5 cm x 8 cm
FormatoPila de transferencia
Longitud (métrico)13,5 cm
MaterialNitrocelulosa
Tamaño de poro0,2 μm
Línea de productosiBlot
Suficiente para10 transferencias
Anchura (métrico)8 cm
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente.

Preguntas frecuentes

If a green color appears on the membrane after the transfer using the iBlot 2 Dry Blotting System, will it affect the results?

The green discoloration is copper deposits from the transfer stack, and it does not affect the results. To minimize this effect, shake excess water off the filter paper and buffer from the gel before placing each on the stack. The current formulation of stacks minimizes the green discoloration.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I cut the bottom stack of my iBlot 2 Transfer Stack to fit my separating gel?

No. Do not trim the iBlot 2 Transfer Stacks to fit your gel size.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Is the PVDF membrane in the iBlot 2 PVDF Gel Transfer Stack pre-activated?

The PVDF membrane is pre-activated and ready for use without any pre-treatment with alcohol.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I use the stack and/or absorbent pad more than once when using the iBlot 2 Dry Blotting System?

No. The transfer stacks have a finite amount of ions to drive the transfer and are depleted after a single use.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I perform multiple transfers, with a new transfer starting immediately after the previous one when using the iBlot 2 Dry Blotting System?

Yes. The device is designed to do so with no impact on performance.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.