iBlot™ Transfer Stack, nitrocellulose, mini, 3 x 10 sets - FAQs
iBlot™ Transfer Stack, nitrocellulose, mini, 3 x 10 sets - FAQs
View additional product information for iBlot™ Transfer Stack, nitrocellulose, mini - FAQs (IB301032)
12 product FAQs found
Following the release of the iBlot 2 Gel Transfer Device, are consumables for the original iBlot Gel Transfer Device still available?
Yes, all original iBlot stacks are still available for purchase. You can find them in the original iBlot Gel Transfer Device Reagents and Resources (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/iblot-dry-blotting-system/reagents-resources-original-iblot-gel-transfer-device.html). However, please note that they are not compatible with the iBlot 2 Gel transfer Device.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is the iBlot 2 Gel Transfer Device compatible with original iBlot Transfer Stacks?
No. Do not use iBlot Transfer Stacks in the iBlot 2 Gel Transfer Device, and do not mix components between iBlot Transfer Stacks and iBlot 2 Transfer Stacks.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How can I get better transfer of high-molecular weight proteins?
Proteins larger than ~150 kDa migrate more slowly than smaller proteins. Therefore, more time is required to transfer them from gel to membrane. We recommend extending the transfer time by 8 to 10 minutes for optimal transfer of proteins >150 kDa using the iBlot Dry Blotting System.
To enhance transfer efficiency, we also recommend adding an equilibration (gel-soaking) step between electrophoresis and western transfer and using NuPAGE Invitrogen 3 - 8% Tris-acetate gels for electrophoresis. We have an application note available, titled “Transferring Large and Small Proteins Using the iBlot Dry Blotting System”. To download the PDF, see the iBlot Reagents Resources page. (http://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/western-blotting/western-blot-transfer/iblot-dry-blotting-system/original-iblot-resources.html)
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Occasionally my western blots have high background. What do you recommend?
This may be a result of insufficient blocking or nonspecific binding. We suggest trying our WesternBreeze Blocker/Diluent (Cat. No. WB7050). We have been using it with good success. Additionally, you should optimize primary and secondary antibody concentrations as generally recommended for any new blotting technique. Many cases of high background can be resolved by further diluting one or both antibody preparations.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is it possible to substitute the membrane from the iBlot Transfer Stack with my specialized membrane?
In theory, you can replace the membrane provided in your iBlot Transfer Stack with any membrane that is compatible with western blotting. To do this, cut the alternative membrane to match the size of your gel, and wet the membrane. Then, either place the alternative membrane on top of the integrated membrane, or carefully remove the integrated membrane from the gel matrix with forceps and replace it with the new membrane. Note that we only support the use of iBlot Transfer Stacks when they are used with the provided instructions.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the shelf life of nitrocellulose and PVDF iBlot Transfer Stacks?
The minimum guaranteed shelf life of iBlot Transfer Stacks is 2 months. Depending on when you purchase the transfer stack, shelf life will be 2–8 months. The expiration date is printed on the package for each stack.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Do the PVDF iBlot Transfer Stacks (0.2 µm, non-autofluorescing) require activation prior to use?
No. The PVDF membrane comes preactivated. You just need to open the transfer stack with membrane, place the separation gel on top of the membrane, and apply one layer of moistened filter paper to run (the same as with the nitrocellulose stacks).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Sometimes there is green discoloration on the blot around the gel when using the iBlot Dry Blotting System. What is this, and does it affect the results?
The green discoloration is copper deposits from the iBlot Transfer Stacks, and it does not affect the results. To minimize this effect, shake excess water off the filter paper and buffer from the gel before placing each on the stack. The current formulation of stacks minimizes the green discoloration.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How can we be more environmentally responsible in regards to the packaging for the iBlot Dry Blotting System?
The plastic in the iBlot Dry Blotting System stack packaging is polyethylene terephthalate (PET) and can be recycled.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I want to conduct western transfer with mini gels (8 x 8 cm), but I do not have iBlot Transfer Stacks (Mini). Can I use iBlot Transfer Stacks (Regular) to transfer my mini gels?
It is best not to transfer a single mini gel on a regular-sized transfer stack. Although in most cases the transfer will be fine, empty spaces on the transfer stack that are not in direct contact with a gel could potentially cause distortions across the whole surface of the membrane, including the portion in contact with the gel. It is best to have more than 50% of a membrane in contact with the gel, if possible.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is there a limitation on the thickness of gels that can be used with the iBlot Dry Blotting System?
The iBlot Dry Blotting System has been tested to efficiently transfer protein from gels ranging in thickness from 1 mm to 3 mm. We have not tested gels thicker than 3 mm because they are rarely used for SDS-PAGE.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.