iBlot™ 3 Starter Kit, PVDF, 1 kit - FAQs
iBlot™ 3 Starter Kit, PVDF, 1 kit - FAQs
View additional product information for iBlot™ 3 Starter Kit, PVDF - FAQs (A56729, IB31002S)
1 product FAQs found
Do you offer additional years of service for the iBlot 3 Western Transfer Device?
No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.