iBlot™ 3 Starter Kit, low-fluorescence PVDF
iBlot™ 3 Starter Kit, low-fluorescence PVDF
iBlot™ 3 Starter Kit, low-fluorescence PVDF
Invitrogen™
iBlot™ 3 Starter Kit, low-fluorescence PVDF
The iBlot 3 Starter Kit, Low Fluorescence PVDF, includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, Low Fluorescence PVDF—everything needed to perform a rapid dry-transfer in a fluorescent western blotting experiment.
Have Questions?
Catalog NumberQuantity
IB31003S1 Kit
Catalog number IB31003S
Price (USD)
5,020.00
Each
Quantity:
1 Kit
Price (USD)
5,020.00
Each
The iBlot 3 Starter Kit, Low Fluorescence PVDF, includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, Low Fluorescence PVDF—everything needed to perform a rapid dry-transfer of proteins from a gel to a membrane in a fluorescent western blotting experiment.
iBlot 3 Western Blot Transfer Device
The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose, polyvinylidene difluoride (PVDF), and Low Fluorescence PVDF membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time.
The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and pre-assembled transfer stacks that contain the required buffers and transfer membrane (nitrocellulose, PVDF, or Low Fluorescence PVDF).
iBlot 3 Western Blot Transfer Device features include:
  • Complete protein transfer in as few as three minutes
  • High detection sensitivity and even, consistent protein transfer
  • Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
  • Built-in adjustable cooling to help ensure consistent transfer temperature
  • Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
  • Ability to create custom programs
  • A simple, user-friendly interface
How the iBlot dry blotting system works
Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion. (Conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen.)
With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dimensions (L x W x H)44 x 30 x 18.25 cm
Frequency50/60 Hz
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferDry
Quantity1 Kit
Throughput2 Midi or 4 Mini Gels
WarrantyStandard, 1-year warranty included
Weight (Metric)8.3 kg
Capacity2 Midi-Gels, 4 Mini-Gels
For Use With (Application)Western Blot
For Use With (Equipment)iBlot 3
Gel CompatibilityMini- or Midi-sized Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, Tris-Glycine Gels
Gel SizeMidi
Product LineiBlot 3
Running Time∼3 to 8 min.
TypeDry Transfer System
Unit SizeEach
Contents & Storage
iBlot 3 Western Blot Transfer Device; Western Blot Roller; iBlot 3 Transfer Stacks, midi, Low Fluorescence PVDF (2 boxes, 20 stacks)
Frequently asked questions (FAQs)
Can I use the low fluorescence PDVF membranes that are provided with the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) in a chemiluminescent western blot?
Yes, the low fluorescence PVDF membrane included with the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) performs similarly to standard PVDF in chemiluminescent applications.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why aren't the membranes in the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) pre-activated?
To minimize membrane autofluorescence, the PVDF membranes are packaged dry. Hence, an activation step must be performed prior to use. If autofluorescence is not a concern, we recommend using iBlot 3 Transfer Stacks, PVDF (Cat. No. IB34001 (midi) or Cat. No. IB34002 (mini)) which contain pre-activated PVDF membranes already incorporated into the transfer stack.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
In the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)), why is the membrane packaged outside of the transfer stack?
To minimize membrane autofluorescence, the PVDF membranes are packaged dry. Hence, an activation step must be performed prior to use. If autofluorescence is not a concern, we recommend using iBlot 3 Transfer Stacks, PVDF (Cat. No. IB34001 (midi) or Cat. No. IB34002 (mini)) which contain pre-activated PVDF membranes already incorporated into the transfer stack.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How long should I activate PVDF membrane in methanol?
We recommend wetting PVDF in 100% methanol or ethanol for 3 min and then rinsing with deionized (DI) water before use. Wetting for shorter times can result in incomplete activation, leading to inconsistent protein binding. Wetting for a longer time will not have a negative impact.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why does PVDF need to be activated?
PVDF is a hydrophobic membrane that will not readily interact or wet in water. To allow protein transfer and binding, the membrane must be initially wetted in methanol or ethanol and then rinsed with deionized (DI) water to allow protein binding to occur during transfer.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.