iBlot™ 3 Transfer Stacks, midi, low-fluorescence PVDF
iBlot™ 3 Transfer Stacks, midi, low-fluorescence PVDF
iBlot™ 3 Transfer Stacks, midi, low-fluorescence PVDF
Invitrogen™

iBlot™ 3 Transfer Stacks, midi, low-fluorescence PVDF

Invitrogen Low Fluorescence PVDF Membranes exhibit exceptionally low autofluorescence across all commonly used fluorescent imaging channels. Low fluorescent background enhances signal-to-noise ratios, resulting in more sensitive, higher quality and more reliable western blot data.
Have Questions?
Catalog NumberQuantity
IB3400310 Stacks
Catalog number IB34003
Price (USD)
335.00
Each
Quantity:
10 Stacks
Price (USD)
335.00
Each

Invitrogen iBlot 3 Transfer Stacks are used with the iBlot 3 Western Blot Transfer Device to transfer proteins from an SDS-PAGE gel to a membrane when performing western blotting experiments. These pre-assembled, single-use consumables utilize copper-fiber electrode sheets and optimized cathode and anode solid buffer gels to eliminate several sources of variability—helping ensure consistent and repeatable results compared with classical methods.

The Low Fluorescence PVDF membranes exhibit exceptionally low autofluorescence across all commonly used fluorescent imaging channels. Low fluorescent background enhances signal-to-noise ratios, resulting in more sensitive, higher quality, and more reliable western blot data.

iBlot 3 Transfer Stacks allow you to:

  • Boost signal-to-noise-ratio—low autofluorescence enhances signal clarity
  • Expand multiplexing capability—low fluorescent background across channels broadens fluorophore options allowing users to detect more targets or minimize spectral cross-talk
  • Minimize variability—single-use copper electrodes prevent salt build-up from impacting results over time
  • Expedite setup—pre-assembled transfer stacks contain solid gel ion matrix, which replaces traditional transfer buffer
  • Simplify clean up—transfer stack is self-contained within a disposable tray, simplifying cleanup to one step (wiping the blotting surface of the transfer device)

Compatibility

Although optimized for fluorescent detection, Low Fluorescence PVDF membranes perform similarly to standard PVDF when used in chemiluminescent detection.

All iBlot 3 transfer stacks are compatible with common commercially available protein gels, including Bis-Tris, Tris-Glycine, Tris-Acetate, and Tricine chemistries. They are intended for use on the iBlot 3 Transfer Device only.

Use

Midi-size transfer stacks (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels. To enable optimal performance in fluorescent applications, Low Fluorescence PVDF membranes are provided dry, in a separate pouch within the box. Activate the membrane before use by soaking in 100% methanol or ethanol, followed by a quick rinse with distilled water. Once activated, simply assemble the transfer stack, choose a transfer method, and press Go.

The iBlot 3 Western Blot Transfer System

The iBlot 3 Western Blot Transfer System, which includes iBlot 3 Transfer Stacks and the iBlot 3 Western Blot Transfer Device, utilizes dry blotting technology to produce higher quality and more reliable western blot data while reducing protein transfer to 3–8 minutes.

The iBlot 3 system features two independently controlled transfer stations, allowing the device to be shared with coworkers and simultaneously running up to four mini gels or two midi gels with two different programs. In addition, the robust design and adjustable cooling function reduces heat buildup, facilitating better run-to-run consistency.

How dry blotting technology works

Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion. (Conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen.)

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our exceptional de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Western Blotting
For Use With (Equipment)iBlot 3
Quantity10 Stacks
Dimensions (LxW)13.5 x 8 cm (Midi)
FormatTransfer Stack
Length (Metric)13.5 cm
MaterialPVDF
Pore Size0.3 μm
Product LineiBlot
Sufficient For10 Blots
Width (Metric)8 cm
Unit SizeEach
Contents & Storage
10 transfer stacks, 10 Low Fluorescence PVDF Membranes, 10 Absorbent Pads, 10 Filter Papers; Store at room temperature

Frequently asked questions (FAQs)

Can I use the low fluorescence PDVF membranes that are provided with the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) in a chemiluminescent western blot?

Yes, the low fluorescence PVDF membrane included with the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) performs similarly to standard PVDF in chemiluminescent applications.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Why aren't the membranes in the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)) pre-activated?

To minimize membrane autofluorescence, the PVDF membranes are packaged dry. Hence, an activation step must be performed prior to use. If autofluorescence is not a concern, we recommend using iBlot 3 Transfer Stacks, PVDF (Cat. No. IB34001 (midi) or Cat. No. IB34002 (mini)) which contain pre-activated PVDF membranes already incorporated into the transfer stack.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

In the iBlot 3 Transfer Stacks, low fluorescence, PVDF (Cat. No. IB34003 (midi) and Cat. No. IB34004 (mini)), why is the membrane packaged outside of the transfer stack?

To minimize membrane autofluorescence, the PVDF membranes are packaged dry. Hence, an activation step must be performed prior to use. If autofluorescence is not a concern, we recommend using iBlot 3 Transfer Stacks, PVDF (Cat. No. IB34001 (midi) or Cat. No. IB34002 (mini)) which contain pre-activated PVDF membranes already incorporated into the transfer stack.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How long should I activate PVDF membrane in methanol?

We recommend wetting PVDF in 100% methanol or ethanol for 3 min and then rinsing with deionized (DI) water before use. Wetting for shorter times can result in incomplete activation, leading to inconsistent protein binding. Wetting for a longer time will not have a negative impact.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Why does PVDF need to be activated?

PVDF is a hydrophobic membrane that will not readily interact or wet in water. To allow protein transfer and binding, the membrane must be initially wetted in methanol or ethanol and then rinsed with deionized (DI) water to allow protein binding to occur during transfer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.