Dionex ICS-6000 Single Channel Gradient Standard Bore IC System - FAQs
Dionex ICS-6000 Single Channel Gradient Standard Bore IC System - FAQs
View additional product information for Dionex™ ICS-6000 Standard Bore and Microbore HPIC™ Systems - FAQs (ICS6000-007, ICS6000-008, ICS6000-009, ICS6000-013, ICS6000-014, ICS6000-015, ICS6000-010, ICS6000-011, ICS6000-012)
15 product FAQs found
What kind of support will I get for my Dionex ion chromatography system? 
 We provide comprehensive support. Our specialists act as trusted advisors and share industry expertise. They understand that purchasing an IC instrument is also purchasing a relationship, and they strive to provide the level of service and support that laboratories need. 
What validation services do you offer for the Dionex ICS-6000 HPIC System?  
 We offer a full range of validation services and kits for the Dionex ICS-6000 product line, including Installation Qualification (IQ) and Operational Qualification (OQ) procedures. All validation kits include detailed procedures, calibration tools, data worksheets, methods, and validation certificates. 
What are the automated sample preparation capabilities of the Dionex ICS-6000 HPIC System? 
 The Dionex ICS-6000 HPIC system offers the most comprehensive suite of sample preparation features along with the ability to combine them into a single workflow. The system can perform filtration, dilution, matrix elimination, and pre-concentration when paired with our portfolio of autosamplers, in-line and offline cartridges, electrolytic water purifiers (EWPs), and trap and concentrator columns. 
How does the Dionex ICS-6000 HPIC System handle trace analysis? 
 The system is ideal for 2D-IC analysis, which is particularly important when performing trace analysis in the presence of high concentrations of interfering matrix ions. This technique achieves improved selectivity and signal enhancement without complicated automated sample preparation.
What advantages does the Dionex ICS-6000 HPIC system offer for carbohydrate analysis? 
 The system enables direct detection of simple to complex carbohydrates using electrochemical analysis without sample derivatization (HPAE-PAD). The Dionex ICS-6000 HPIC system offers the widest variety of elution techniques for carbohydrate analysis, including proportioned gradients and dual eluent generation. Using Dual EGC mode, the Dionex ICS-6000 HPIC system can also generate potassium hydroxide/potassium methanesulfonate (KOH/KMSA) eluents for simple to complex carbohydrate analysis by adding just deionized water. 
What detection options does the Dionex ICS-6000 HPIC System support? 
The system supports multiple detection options:
  • Suppressed and non-suppressed conductivity
  • Electrochemical (DC amperometry and integrated pulsed amperometry)
  • UV-Vis absorbance (variable wavelength and photodiode array)
  • Inductively coupled plasma mass spectrometry (ICP-MS)
  • Mass spectrometry (MS)
Is the Dionex ICS-6000 HPIC System compatible with mass spectrometry?  
 Yes. The system is compatible with single-quadrupole, triple-quadrupole, and high-resolution accurate-mass (HRAM) mass spectrometry, including Thermo Scientific Orbitrap mass spectrometers, for performing powerful ion chromatography-mass spectrometry (IC-MS) analyses.
Can the Dionex ICS-6000 HPIC System be configured as a dual-channel system? 
 Yes, the system can be easily converted to a dual-channel system, allowing you to run two different analyses simultaneously on a single sample or analyze two different samples concurrently without taking up additional bench space as it can share one autosampler for both channels. 
What column formats does the Dionex ICS-6000 HPIC System support? 
The system supports multiple column formats:
  • Microbore (1–3 mm column i.d.)
  • Standard bore (3–7 mm column i.d.)
What flow rates does the Dionex ICS-6000 HPIC system support? 
The pumps of the Dionex ICS-6000 HPIC System support flow rates from 0.001 to 10.000 mL/min, with specific formats offering:
Microbore: 0.2–0.5 mL/min
  • Standard bore: 1–2 mL/min
  • What pressure capabilities does the Dionex ICS-6000 HPIC System offer?  
    The system can operate at pressures up to 6000 psi (5000 psi for continuous operation) with RFIC technology, allowing for faster analysis with higher flow rates and smaller-particle-size columns without sacrificing resolution.
    What is the Thermo Scientific Dionex ICS-6000 HPIC System? 
    Dionex ICS-6000 HPIC System is a modular, high-pressure ion chromatography (HPIC) system. It is a highly configurable IC system, designed for users who want flexibility to solve complex analysis challenges.
    The system’s high-pressure operation allows you to use smaller-particle-size columns, such as 4 µm particle-size columns for fast analyses and high resolution. Its Reagent-Free IC (RFIC) capabilities provide reproducibility and ease-of-use, and its modular design makes it both adaptable for a wide variety of analyses and upgradable, should you need additional capabilities later.
    What software can be used with the Dionex ion chromatography systems? 
     The system uses the Thermo Scientific Chromeleon Chromatography Data System (CDS), which unifies and simplifies system control, operation, data processing and collection, and reporting. 
    How does the Dionex Integrion HPIC system improve laboratory throughput?  
     By increasing pressure tolerance up to 6000 psi, smaller particle size columns such as 4 µm particle-size columns can be used to achieve shorter run times. This allows the system to provide increased throughput and productivity without sacrificing resolution or data quality. Automated eluent generation provides time savings by eliminating human errors associated with manual eluent preparation. 
    What does HPIC stand for in Thermo Scientific Dionex ion chromatography systems and why is it important?  
     HPIC stands for high-pressure ion chromatography. This capability enables the use of smaller-particle-size 4 µm columns, which provide faster flow rates without sacrificing resolution. HPIC results in shorter run times and increased laboratory throughput while maintaining data quality.