iGlow™ Sample Tray Holder
iGlow™ Sample Tray Holder
iGlow™ Sample Tray Holder
Invitrogen™
iGlow™ Sample Tray Holder
The Invitrogen iGlow Sample Tray Holder organizes storage of iGlow Gel Documentation System accessories. This aluminum stand features slots to accommodate the UV (IGA011), Blue (IGA012), and White (IGA013) sample trays and the iGlow UV Safety Guard (IGA015).
Have Questions?
Catalog NumberQuantity
IGA0141 Ea.
Catalog number IGA014
Price (USD)
90.00
Each
Quantity:
1 Ea.
Price (USD)
90.00
Each
The Invitrogen iGlow Sample Tray Holder organizes storage of iGlow Gel Documentation System accessories. This aluminum stand features slots to accommodate the UV (IGA011), Blue (IGA012), and White (IGA013) sample trays and the iGlow UV Safety Guard (IGA015). The robust construction enables secure storage of these components in a visible and convenient location, facilitating efficient lab work and protecting valuable accessories from damage or misplacement.
Tray Holder Features and Specifications
The iGlow tray holder is made with shaped and treated aluminum. This enables consistent performance even under heavy usage in laboratory conditions.
The compact design of the tray holder enables storage of all iGlow accessories in a small space, preserving bench space while protecting components from damage or misplacement. This upright configuration also enables high visibility of the sample trays and helps ensure easy handling and storage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorWhite
For Use With (Application)DNA and Visible Gel Imaging
For Use With (Equipment)iGlow Gel Documentation System
IncludesStorage space for 4 imaging accessories
Product LineiGlow
Quantity1 Ea.
Product TypeGel Documentation Kit
Unit SizeEach
Frequently asked questions (FAQs)
What maintenance is required for the iGlow Gel Documentation System and imaging accessories?
No routine calibration or replacement parts are required. When needed, the sample trays, touchscreen, and transilluminator glass may be cleaned using deionized water, 70% ethanol, or isopropyl alcohol followed by wiping with a lint-free cloth or tissue.
Are replacement imaging trays for the iGlow Gel Documentation System available?
Yes. Replacement imaging trays are available for purchase. The UV imaging tray is Cat. No. IGA011, the blue imaging tray is Cat. No. IGA012, and the white imaging tray is Cat. No. IGA013.
Is the iGlow sample tray holder compatible with iBright imaging accessories?
Yes. The iGlow sample tray holder is compatible with select iBright imaging accessories, including the contrast tray and glass turntable.
What accessories are included with the iGlow Gel Documentation System?
The iGlow Gel Documentation System includes the UV, blue, and white trays required for imaging, the UV Safety Guard for gel excision, and the sample tray holder.
Are the iGlow imaging trays compatible with other instrumentation?
No. The imaging trays are designed specifically for use with the iGlow Gel Documentation System.