LInspector Edge In-line Mass Profilometer
LInspector Edge In-line Mass Profilometer
LInspector Edge In-line Mass Profilometer
LInspector Edge In-line Mass Profilometer
Thermo Scientific™
LInspector Edge In-line Mass Profilometer
The Thermo Scientific LInspector Edge In-line Mass Profilometer harnesses the new analytical technique of mass profilometry to deliver enhanced electrode quality assurance.
Have Questions?
Catalog NumberApplications
IMP-110800-1Lithium-Ion Batteries
Catalog number IMP-110800-1
Price (USD)
-
Applications:
Lithium-Ion Batteries
To make higher performing, safer batteries at competitive costs, battery manufacturers need to meet increasingly demanding levels of electrode coating uniformity. Thermo Scientific LInspector Edge In-line Mass Profilometer harnesses the new analytical technique of mass profilometry to deliver significantly enhanced electrode quality assurance. Measuring complete edge-to-edge coating profiles in milliseconds, with exemplary resolution and precision, it inspects the entire electrode to provide full traceability for each electrode patch, stripe, or roll. Providing previously inaccessible levels of information and insight, the LInspector Edge In-line Mass Profilometer enables the responsive production control required to minimize transition times and scrap, while maximizing quality and yield.
Battery manufacturing with unprecedented coating QA
The Thermo Scientific LInspector Edge In-line Mass Profilometer is a next-generation process metrology tool for battery manufacturing that sets a new benchmark for electrode coating QA. It measures 100% of a coated electrode in real time, without scanning and with no moving parts. Both full-width basis weight and profile uniformity analysis are provided by simultaneously gathering spatial information and coating weight data. These capabilities are complemented by unprecedented precision and the ability to capture data at full production speeds, producing high resolution images, rather than data from the spot measurements produced by conventional basis weight gauges traversing across the sheet width. The net result is orders of magnitude more coating profile weight data – as much as 1000X more than conventional gauges generate – along with detection of non-uniformities that other technologies miss. With the LInspector Edge In-line Mass Profilometer, battery manufacturers have the most secure foundation for exemplary, responsive, and accurate process control and the ability to safeguard the very highest levels of quality and performance.
  • 100% electrode mass loading inspection
  • Responsive, advanced process control
  • Complete traceability for quality assurance
100% electrode mass loading inspection
The LInspector Edge In-line Mass Profilometer allows battery manufacturers to take active control of their electrode coating quality. Capable of operating at high production speeds, it measures 100% of the coated electrode in real time, delivering 1000X more mass loading uniformity data than traditional scanning gauges. This wealth of information allows users to catch small defects early in the process in time for effective remedial action, benefitting battery manufacturers in multiple ways:
  • Reduced scrap rates
  • Reduced time to market
  • Improved process understanding and product performance
Responsive, advanced process control
Electrode coating weight and profile uniformity is vital to maintaining the quality and safety of lithium-ion batteries, and the LInspector Edge In-line Mass Profilometer helps manufacturers to meet specifications while maximizing process efficiency. It delivers a full coating profile every 3 millimeters, providing the fastest measurement feedback for quicker, better-informed, and more accurate control decisions. This enables swifter start-up and accelerated product transitions to reduce scrap and increase throughput.
Complete traceability for quality assurance
The quality of battery components must be maintained throughout the manufacturing process; even small defects can have serious implications for the safety and performance of the battery. Therefore, battery manufacturers need to implement a system of complete traceability that allows them to identify defects early in the process and trace them back to the source.
Traceability of an entire electrode roll is finally possible with the LInspector Edge In-line Mass Profilometer through complete coating uniformity measurements for the entire production run, in the form of easily accessible, high-resolution images. This new mass profilometry technology can deliver visibility of the entire life cycle of every electrode roll with valuable data to quickly spot production issues or trends. In doing so, it allows battery manufacturers to make proactive improvements to optimize the production process for maximum efficiency.
Specifications
DescriptionElectrode coating
ApplicationsLithium-Ion Batteries
Unit SizeEach