InSight Kits
Actual product may vary
InSight Kits
InSight Kits
InSight Kits
InSight Kits
InSight Kits
InSight Kits
Thermo Scientific™
InSight Kits
Proactively monitor your cold storage equipment’s temperature, energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
INSKTCRYO101LN2 Cryo Storage
KITT3C1D2WIF1001ULT Freezers
INSKTCOLD101ULT Freezers, Freezers, Refrigerators
KITT3C1D2WIF1002Refrigerators
INSKTWALK101Walk-in w/ Single Refrigeration System
INSKTWALK102Walk-in w/ Dual Refrigeration System
Catalog number INSKTCRYO101
Price (USD)
-
For Use With (Equipment):
LN2 Cryo Storage
Thermo Scientific™ InSight is a wireless monitoring solution that has been designed to proactively monitor your cold storage equipment. InSight consists of wireless modules (motes), sensors and software tools that allow you to monitor your cold storage equipment’s temperature, energy consumption, door access, reliability, predictive failure analytics, as well as other parameters using Wi-Fi or proprietary wireless protocols. Alarms are issued automatically to users’ email and mobile devices when setpoint breaches are detected in the system.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Cabinet temperature
  • Electrical use and energy consumption
  • Door openings
  • Ambient temperature
InSight Analytics monitors your cold storage equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight gateway server. There, the server collects and transmits the data to Cloud storage.
  • As an alternative to Wi-Fi, wireless 915MHZ systems (North America only) are also available for smaller installations (25 motes or less).
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the cold storage equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
Not available outside North America
Specifications
CalibrationNot Calibrated
DescriptionInSight Kit for Cryogenic Storage (LN2)
For Use With (Equipment)LN2 Cryo Storage
Alarm TypesMobile Device
Display TypeNo Display
Software TypeInsightlink Gateway
TypeInsight Monitoring Kit
Unit SizeEach