Qtegra™ ISDS Software for Trace Elemental Analysis - FAQs
Qtegra™ ISDS Software for Trace Elemental Analysis - FAQs
View additional product information for Qtegra™ Intelligent Scientific Data Solution™ Software - FAQs (IQLAAGGACBFAOVMBJD, IQLAAMGAEFFAOVMBCZ, BRE0032411, IQLAAGGACBFAOVMBCZ, BRE0064051)
11 product FAQs found
Can I run my IC for speciation from inside the Qtegra ISDS software, or do I still have to juggle two programs?
Yes—add the ChromControl option and control the chromatograph from the same screen as the mass spectrometer. Sequences, timing, and reporting stay synchronized so your speciation runs feel like a single system.
Does Qtegra ISDS Software contain solutions based on Artificial Intelligence/Machine Learning (AI/ML)?
Qtegra ISDS Software provides a series of tools to support analysts in their daily tasks. For example, when using a Thermo Scientific iCAP PRO Series ICP-OES system, the SemiQuant feature within the Qtegra ISDS Software includes a data acquisition tool that leverages Artificial Intelligence and neural networks to simplify the complex process of intra- and inter-element correction, automatically selecting only non-interfered peaks. SemiQuant supports fast screening of samples to provide rapid overviews of sample composition without requiring any calibration.
For trace level work, can Qtegra ISDS software build the full QA/QC sequence for me and flag results that are out of range?
Templates include comprehensive QC tests with automatic pass/fail actions, and the software highlights results that exceed your set limits, so low level work stays disciplined without extra clicks.
Is Qtegra ISDS Software 21 CFR Part 11 compliant?
Although no software package can be said to be compliant with respect to 21 CFR Part 11, Qtegra ISDS Software provides the required tools to implement the correct (procedural, administrative and technical) controls to be compliant with 21 CFR Part 11 for ICP-OES, Quadrupole ICP-MS and Gas IRMS systems. It includes tools to facilitate access control, generate different user levels, create audit trails and support for electronic signatures. All these features are an integral part of Qtegra ISDS Software and do not require additional software packages to be installed.
How does the Qtegra ISDS Software integrate with Laboratory Information Management Systems (LIMS)?
The Qtegra ISDS system is designed for seamless, universal integration with LIMS systems. Sample information can be taken into the analytical workflow in various ways, for example through external data files, or from (bar or QR) digital codes from samples presented for analysis. Data can be automatically reported (using flexible, user-defined templates), exported or printed, either directly after each sample is analyzed or as a complete data set.
Can Qtegra ISDS software send results straight to spreadsheets or a LIMS without copy paste?
Yes. Results can be pushed to files you define for spreadsheets or forwarded to your data system, and you can build report layouts so analysts don’t need to edit outputs by hand.
Can Qtegra ISDS software automatically dilute over range samples so I don’t have to rerun them by hand?
Qtegra ISDS software connects to compatible autodilution hardware and applies rules to re measure off scale samples automatically. That means fewer manual interventions and more consistent recoveries across a wide concentration range.
If we add another analyzer later, can we keep the same Qtegra ISDS software interface or will my team have to learn something new?
You keep the same interface. Qtegra ISDS software covers ICP OES, ICP MS, and multicollector ICP-MS as well as isotope ratio instruments, so your team can move between techniques without switching software or retraining.
We work in regulated labs. Can Qtegra ISDS Software handle controlled access and formal review/approval without a lot of IT effort?
Yes. Qtegra ISDS Software visually flags results that fall outside user-defined criteria, simplifying data review. It includes instrument monitoring for consistent performance in accredited laboratories and offers features for method validation, data security, and complete ‘who, what, when, why’ traceability to meet the requirements of a GxP-compliant laboratory. The software includes role based permissions, time stamped activity logs, and built in review/approve steps. Records are encrypted and the system can be configured for closed or open environments, so you can meet governance needs with tools already in the box.
How does Qtegra ISDS Software improve laboratory productivity?
Qtegra ISDS Software contains a series of functions to support analysts through automation of tasks in their routine workflow (for specific instruments only). One example is Get Ready, which automates plasma start and performance verification of the system at user defined times for Thermo Scientific iCAP MX Series ICP-MS systems. The iCAP MX Series ICP-MS systems also include the HAWK Consumable and Maintenance Assistant, which notifies analysts when maintenance is required. The Step Ahead function allows analysts to overlap data acquisition with rinsing of the autosampler needle, increasing sample throughput by up to 25% when using the Thermo Scientific iSC-65 autosampler. For isotope analysis using Thermo Scientific DELTA Q IRMS, Qtegra ISDS Software supports seamless switching between different peripherals (e.g., EA and GC), avoiding downtime and manual interventions. For Thermo Scientific GasBench Plus IRMS System, Qtegra ISDS Software allows for automated sampling and sample preparation switching between trays, increasing the number of samples that can be processed.
How will Qtegra ISDS Software make my day to day runs simpler?
Qtegra ISDS Software is instrument-agnostic software that offers a single platform for both elemental and isotope analysis. This software offers the same workflow, functionality and look and feel across multiple instruments. Analysts can flexibly switch between different techniques, eliminating the hassle of having to learn a completely new software package when, for example, introducing new instruments to the laboratory. It offers one click startup, a guided path from sample to result, and a LabBook that keeps methods, sequences, results, and QC together. Because the same interface runs across techniques, setup takes fewer steps and reporting is faster.