Can I run my IC for speciation from inside the Qtegra ISDS software, or do I still have to juggle two programs?
Yes—add the ChromControl option and control the chromatograph from the same screen as the mass spectrometer. Sequences, timing, and reporting stay synchronized so your speciation runs feel like a single system.