ARL X900  Simultaneous/Sequential XRF Spectrometer
ARL X900  Simultaneous/Sequential XRF Spectrometer
ARL X900  Simultaneous/Sequential XRF Spectrometer
Thermo Scientific™
ARL X900 Simultaneous/Sequential XRF Spectrometer
The Thermo Scientific™ ARL™ X900 is a robust WDXRF analyzer with XRD capability that provides quick, precise elemental analysis for industrial process control.
Have Questions?
Catalog NumberType
IQLAAHGABMFARVMBNQSimultaneous-Sequential XRF Spectrometer
Catalog number IQLAAHGABMFARVMBNQ
Price (MXN)
-
Type:
Simultaneous-Sequential XRF Spectrometer
The Thermo Scientific™ ARL™ X900 XRF Spectrometer is a robust WDXRF analyzer with XRD capability that provides quick, precise elemental composition results, thanks to dual loading of samples and fast analysis. With a reliable design to maximize productivity, the ARL X900 Spectrometer can keep up with modern metals, cement and mining production and process control needs.
Elemental analysis is a cornerstone of the mining, cement, and metals industries. The ability to quickly and accurately assess the elements present in raw materials, products, and by-products is essential to those industries' ability to ensure the quality of their products and their process. The ARL X900 sequential/simultaneous wavelength-dispersive XRF spectrometer is most suitable for XRF elemental analysis and process control. It delivers reliable and precise elemental analysis results quickly, whatever your process control needs. Maximum reliability, and durability is obtained through a careful and robust design, engineered to handle demanding industrial settings. The analytical core of the ARL X900 Spectrometer, is protected inside a vacuum chamber that ensures very stable analytical conditions. The 'tube above sample' geometry is inherently safe. This means fewer unplanned interventions, and excellent short- and long-term stability, which is critical for good analytical results.
Application domains:
Metals: cast irons, steels, slags, ferro-alloys, direct reduced iron; copper, brass and bronze alloys; aluminum smelting (bath ratio) and aluminum alloys; zinc alloys, tin alloys, magnesium alloys, nickel alloys, super-alloys, titanium alloys, cobalt alloys
Cement: raw meal, clinker, slags, limestone, free lime and clinker phases, etc.
Mining & minerals: rocks, ores, concentrators, etc.
Others: ceramics, glasses, polymers, contract laboratories
Features:
  • Combined simultaneous and sequential WDXRF capabilities, bringing unrivalled analytical flexibility and speed
  • Able to perform elemental analysis with concentrations ranging from parts per million to 100%
    • Flexibility with sequential analysis from B to Am with goniometer
    • Ultra-fast simultaneous analysis with fixed channel monochromators
  • Compact XRD for additional crystalline phase analysis provides two techniques in one instrument
  • Durable and robust instrument design with tube above geometry and a single analysis chamber imparts maximum reliability
  • 'LoVap technology' X-ray tube ensures excellent long-term stability and outstanding analytical performance
  • Fast dual-sample loader optimizes throughput by reducing dead time between measurements.
  • Factory calibrations enable immediate use for a wide range of applications
  • UniQuant Standardless analysis provides analysis results of unknown samples within minutes
  • OXSAS Analytical Software includes all the features needed for data management, instrument control, calibration, instrument set-up and diagnostics with wide analytical capacity and flexibility
  • Optional automation capabilities allow automatic, unattended round-the-clock operation
Efficiency and flexibility
Faster analysis and increased throughput are achieved with the dual-sample loader that reduces dead time between samples: During sample analysis, the next sample is being loaded. Further time savings are achieved by combining simultaneous analysis and sequential analysis, working in parallel, in one spectrometer. The system is configurable with up to 32 fixed channels, up to 2 goniometers, and a compact XRD system.
Automation and software
Automation is available to increase sample throughput, improve repeatability, and allow operations to meet tighter product specifications and time pressures without increasing overhead costs. All automation solutions are customized to meet your specific requirements.
The ARL X900 Spectrometer uses Thermo Scientific™ OXSAS™ Analytical Software. This simple, flexible, and powerful software is easy to use, no matter what the requirements are. From simple tasks to complex jobs, OXSAS Software allows them to be defined easily and performed quickly. Automated control makes unattended analysis possible, while programming can ensure support of priority samples.
Several features on the ARL X900 Spectrometer enhance process control in multiple ways:
  • Simple EasyOXSAS interface for streamlined routine analysis
  • Numerous customizable on-line processing functions, with manual or automatic application
  • Real on-line integrated statistical process control (SPC)
  • SCT Manager provides status and history of setting-up samples, control samples, and type standards, thus giving an overview of the analytical capability of the instrument and of each method at any time
Analysis you can depend on
The ARL X900 XRF Spectrometer delivers reliability and robustness. With Thermo Fisher Scientific’s support and service you can count on, the ARL X900 Spectrometer is set to be the trusted standard in elemental analysis. Its configuration can be customized, and calibrations tailored to fit your industrial processes, so that you can focus on product quality and process efficiency.
Specifications
DescriptionWDXRF analyzer with XRD capability for quick, precise elemental analysis for the minerals and mining, cement and metals industries to enable industrial process controls
IncludesOXSAS™ Analytical Software
TypeSimultaneous-Sequential XRF Spectrometer
Unit SizeEach
Frequently asked questions (FAQs)
What advantages does integrated XRF and XRD analysis provide?
Integrated XRF and XRD analysis enables laboratories to obtain both elemental composition and crystalline phase information from a single instrument platform. XRF determines which elements are present and their concentrations, while XRD identifies the crystalline phases and mineralogical composition of a sample. Combining these techniques can help streamline workflows, no external sample transfer required, reduce the need for multiple instruments, and provide a more complete understanding of materials for applications such as cement production, mining, metals processing, and industrial process control.
Can the ARL X900 analyze unknown materials without calibration standards?
Yes. The Thermo Scientific ARL X900 can support standardless analysis using Thermo Scientific UniQuant software. This capability can help users identify and quantify elemental composition in unknown materials when application-appropriate calibration standards are unavailable.
Can the ARL X900 support high-throughput and unattended operation?
Yes. The Thermo Scientific ARL X900 includes features such as dual-sample loading, automation options, and OXSAS analytical software to support high-throughput analytical workflows. Automated operation can help laboratories manage routine analyses and prioritize critical samples while maintaining efficient instrument utilization.
How does the ARL X900 combine XRF and XRD analysis?
The Thermo Scientific ARL X900 can be configured with integrated X-ray diffraction (XRD) capability in addition to WDXRF elemental analysis. This enables laboratories to perform elemental composition analysis and crystalline phase analysis using a single instrument platform, which can help streamline workflows for applications such as cement production, mining, and metallurgical process control.
Which industries and laboratories are best suited for the ARL X900 XRF Spectrometer?
The Thermo Scientific ARL X900 is well suited for mining, metals, cement, mineral processing, industrial manufacturing, and contract laboratories that require reliable elemental analysis and process control capabilities. It is designed for environments where routine analysis, high sample throughput, and operational efficiency are critical.