iPrep™ gDNA Forensic Card - FAQs
iPrep™ gDNA Forensic Card - FAQs
View additional product information for iPrep™ gDNA Forensic Card - FAQs (IS10011)
1 product FAQs found
Do you still offer the iPrep Purification Instrument?
The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.