Product Image
Invitrogen™

iPrep™ Card: Total RNA

iPrep™ Card: Total RNA
Have Questions?
Catalog NumberQuantity
IS100141 Card
Catalog number IS10014
Price (USD)
1,166.00
Each
Quantity:
1 Card
Price (USD)
1,166.00
Each

The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)iPrep™
Product LineiPrep
Quantity1 Card
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Do you still offer the iPrep Purification Instrument?

The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.