Do you still offer the iPrep Purification Instrument?
The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.
Search
View additional product information for iPrep™ Cartridge Rack - FAQs (IS10102)
1 product FAQs found