Isotemp™ -86°C Ultra-Low Temperature Freezers, 17.3 cu. ft., Capacity: 17.3 cu. ft. (489.9L); Holds 320 boxes; 208-230V/60Hz; 12A - FAQs
Isotemp™ -86°C Ultra-Low Temperature Freezers, 17.3 cu. ft., Capacity: 17.3 cu. ft. (489.9L); Holds 320 boxes; 208-230V/60Hz; 12A - FAQs
View additional product information for Isotemp™ -86°C Ultra-Low Temperature Freezers, 17.3 cu. ft. - FAQs (IU1786D)