Is BSA present in the Anza Buffers?
As an added convenience, BSA is included in the Anza 10X Buffers and Anza 10X Red Buffers. This eliminates the need to add BSA in a separate step.
Search
View additional product information for Anza™ Buffer Set - FAQs (IVGN2008)
2 product FAQs found