Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
J10662720 L
44497916 x 1 L
A2490410 L
44886211 L
Catalog number J106627
Price (USD)
193.00
Each
Quantity:
20 L
Price (USD)
193.00
Each
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.
Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.
Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.
High Quality
Each batch of Attune™ Focusing Fluid is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Attune™ Acoustic Focusing Cytometer
FormatLiquid
Product LineAttune
Quantity20 L
Product TypeFocusing Fluid
Unit SizeEach