Attune™ Shutdown Solution (1X)

Attune™ Shutdown Solution (1X)

Attune™ Shutdown Solution is a cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic FocusingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
J1066281 L
A24975250 mL
Catalog number J106628
Price (USD)
181.65
Online exclusive
189.00
Save 7.35
Each
Quantity:
1 L
Price (USD)
181.65
Online exclusive
189.00
Save 7.35
Each
Attune™ Shutdown Solution is a cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer.

Minimizes Bubble Formation
This azide-free shutdown solution has been formulated to minimize bubble formulation that can occur in the system lines overnight, to help keep the fluid lines clean, and to minimize microbial growth in the fluid lines.

Use for Instrument Maintenance
The Shutdown cycle is used at the end of each day to ensure that all sample fluids and dyes have been removed from the system, and that the fluidic lines and pumps are filled with Attune™ Shutdown Solution to prevent salt deposits from forming in the lines. Shutdown takes approximately 20 minutes, but most of the steps are automatic and under the control of the Attune™ Cytometric Software. Shutdown procedures should be run at least once a day, even if the instrument is intended for continuous use. Proper cleaning of the instrument ensures its consistent and accurate operation.

Convenient
The ready-to-use 1X shutdown solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system.

High Quality
The Attune™ Shutdown Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAttune
Quantity1 L
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeShutdown Solution
Unit SizeEach
Contents & Storage
Store at room temperature.