Crystallized Bovine Albumin
Thermo Scientific Chemicals

Crystallized Bovine Albumin

Crystallized Bovine Albumin
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
100 g
Catalog number J10856.22
Price (USD)
1,255.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
1,255.00
Each
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
Trace MetalSodium (Na): 0 - 5.0 mg/gm
Assay (unspecified)(electrophoresis): ≥99%
Assay (unspecified)Protein: ≥ 98%
CommentSource is Bovine blood
Identification (FTIR)Conforms
View more
  • Source: Bovine blood
  • Assay (electrophoresis): ≥99%
  • Identity: By IR
  • Protein: ≥98%
  • pH (7%, 0.15M NaCl): 5.0 - 5.4
  • Moisture (KF) when packaged: %le;5.0%
  • Sodium (Na): 0 - 5.0mg/g
  • Chloride (Cl): 0 - 3.0mg/g
  • CAS: 9048-46-8
RUO – Research Use Only