Enzymatic Digest Casein Hydrolysate
Thermo Scientific Chemicals

Enzymatic Digest Casein Hydrolysate

Enzymatic digest of casein.
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.2 kg (5 lb.)
453.5 g (1 lb.)
11.3 kg (25 lb.)
Catalog number J12855.P5
Price (USD)
306.00
Each
Quantity:
2.2 kg (5 lb.)
Request bulk or custom format
Price (USD)
306.00
Each
Chemical Identifiers
CAS65072-00-6
IUPAC NameCasein hydrolysate
SMILES*
MDL NumberMFCD00130738
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
AssayProtein >90%
Assay (unspecified)Protein >90%
Ash≤ 6.5%
View more
  • Synonym: Peptones
  • pH: 6.6 to 7.5
  • CAS: 65072-00-6
  • MDLNumber: MFCD00282824
RUO – Research Use Only