Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure
Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure
Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure
Thermo Scientific Chemicals

Cholic Acid Sodium Salt, ≥97% (dry basis), Ultrapure

CAS: 361-09-1 | C24H39NaO5 | 430.561 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number J13630.09
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Chemical Identifiers
CAS361-09-1
IUPAC Namesodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Molecular FormulaC24H39NaO5
InChI KeyNRHMKIHPTBHXPF-TUJRSCDTSA-M
SMILES[Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormWhite powder
Assay (unspecified)≥ 97.0% (dry basis)
Loss on Drying≤ 5.0%
RUO – Research Use Only